The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130446.2 | NP_569713.2 | 6 - 7 | 1624 | 99 |
XM_011513273.2 | XP_011511575.1 | 5 - 6 | 1279 | 99 | |
GenBank mRNA | AF441792.1 | - | 6 - 7 | 1536 | 99 |
AK024439.2 | - | 2 - 3 | 424 | 99 | |
AK097125.1 | - | 6 - 7 | 1578 | 99 | |
AK314473.1 | - | 6 - 7 | 1578 | 99 | |
BC032348.1 | - | 6 - 7 | 1602 | 99 |