The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024637.4 | NP_078913.3 | 1 - 2 | 202 | 97 |
GenBank mRNA | AF316113.1 | - | 1 - 2 | 45 | 97 |