The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000072.3 | NP_000063.2 | 7 - 8 | 1118 | 91 |
NM_001001547.2 | NP_001001547.1 | 7 - 8 | 1079 | 91 | |
NM_001001548.2 | NP_001001548.1 | 7 - 8 | 1118 | 91 | |
NM_001127443.1 | NP_001120915.1 | 6 - 7 | 824 | 91 | |
NM_001127444.1 | NP_001120916.1 | 5 - 6 | 999 | 91 | |
NM_001289909.1 | NP_001276838.1 | 5 - 6 | 703 | 91 | |
NM_001289911.1 | NP_001276840.1 | 5 - 6 | 757 | 91 | |
NR_110501.1 | 6 - 7 | 883 | 91 | ||
XM_005250713.1 | XP_005250770.1 | 7 - 8 | 1060 | 91 | |
XM_005250714.1 | XP_005250771.1 | 7 - 8 | 1027 | 91 | |
XM_005250715.4 | XP_005250772.1 | 7 - 8 | 1185 | 91 | |
GenBank mRNA | AK096858.1 | - | 10 - 11 | 1388 | 91 |
AK129899.1 | - | 7 - 8 | 1060 | 91 | |
AK300101.1 | - | 5 - 6 | 757 | 91 | |
AK312920.1 | - | 7 - 8 | 1042 | 91 | |
BC008406.1 | - | 7 - 8 | 993 | 91 | |
HM217020.1 | - | 5 - 6 | 801 | 91 | |
HM217021.1 | - | 5 - 6 | 801 | 91 | |
HM217022.1 | - | 5 - 6 | 801 | 91 | |
HM217023.1 | - | 4 - 5 | 621 | 91 | |
HM217024.1 | - | 5 - 6 | 801 | 91 | |
HM217025.1 | - | 5 - 6 | 801 | 91 | |
KU177961.1 | - | 5 - 6 | 704 | 91 | |
L06850.1 | - | 6 - 7 | 836 | 91 | |
M24795.1 | - | 7 - 8 | 914 | 91 | |
M98398.1 | - | 7 - 8 | 958 | 91 | |
M98399.1 | - | 7 - 8 | 958 | 91 |