The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267814.1 | NP_001254743.1 | 2 - 3 | 350 | 104 |
XM_005250502.2 | XP_005250559.1 | 2 - 3 | 349 | 104 | |
XM_006716054.2 | XP_006716117.1 | 2 - 3 | 331 | 104 | |
GenBank mRNA | AK128873.1 | - | 2 - 3 | 296 | 104 |
BC000154.2 | - | 2 - 3 | 332 | 104 |