Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- FAM13C
- Entrez Gene ID
-
220965
- Gene Name
- family with sequence similarity 13 member C
- Gene Aliases
-
FAM13C1
- Chromosome Location
-
Chr.10: 59245933 - 59363186 on Build GRCh38
View Full Product Details