The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020443.4 | NP_065176.3 | 3 - 4 | 1572 | 62 |
XM_011510097.1 | XP_011508399.1 | 3 - 4 | 1251 | 62 | |
XM_011510098.1 | XP_011508400.1 | 3 - 4 | 1251 | 62 | |
XM_011510099.1 | XP_011508401.1 | 3 - 4 | 1251 | 62 | |
XM_011510100.1 | XP_011508402.1 | 3 - 4 | 1251 | 62 | |
XM_011510101.1 | XP_011508403.1 | 3 - 4 | 1251 | 62 | |
XM_011510102.1 | XP_011508404.1 | 3 - 4 | 1251 | 62 | |
XM_017002751.1 | XP_016858240.1 | 3 - 4 | 1251 | 62 | |
XM_017002753.1 | XP_016858242.1 | 3 - 4 | 1251 | 62 | |
XM_017002754.1 | XP_016858243.1 | 3 - 4 | 1251 | 62 | |
GenBank mRNA | AB032977.3 | - | 3 - 4 | 905 | 62 |
AJ488101.1 | - | 3 - 4 | 1103 | 62 |