The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006859.3 | NP_006850.2 | 6 - 7 | 697 | 85 |
NM_194451.2 | NP_919433.1 | 6 - 7 | 697 | 85 | |
XM_017007667.1 | XP_016863156.1 | 6 - 7 | 690 | 85 | |
GenBank mRNA | AK026289.1 | - | 6 - 7 | 665 | 85 |
AK292238.1 | - | 6 - 7 | 697 | 85 | |
BC023635.2 | - | 6 - 7 | 642 | 85 |