Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YTHDC1
Entrez Gene ID
91746
Gene Name
YTH domain containing 1
Gene Aliases
YT521, YT521-B
Chromosome Location
Chr.4: 68310387 - 68350106 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031732.2 NP_001026902.1 5 - 6 1331 82
XM_005265706.2 XP_005265763.1 5 - 6 1331 82
XM_005265707.2 XP_005265764.1 5 - 6 1465 82
XM_017008832.1 XP_016864321.1 5 - 6 1465 82
GenBank mRNA AB075846.1 - 2 - 3 237 82
BC041119.1 - 5 - 6 1261 82

View Full Product Details