The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033262.4 | NP_150287.1 | 2 - 3 | 2539 | 135 |
GenBank mRNA | AJ304852.1 | - | 1 - 2 | 1847 | 135 |