The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322271.1 | NP_001309200.1 | 10 - 11 | 1549 | 90 |
NM_001322272.1 | NP_001309201.1 | 11 - 12 | 1914 | 90 | |
NM_017437.2 | NP_059133.1 | 11 - 12 | 1709 | 90 | |
GenBank mRNA | AB037788.1 | - | 5 - 6 | 833 | 90 |
AK001627.2 | - | 11 - 12 | 1686 | 90 | |
AK001912.1 | - | 9 - 10 | 1265 | 90 | |
AK023583.1 | - | 8 - 9 | 1286 | 90 | |
AL442079.1 | - | 4 - 5 | 608 | 90 | |
BC070095.1 | - | 11 - 12 | 1679 | 90 |