Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THBS2
Entrez Gene ID
7058
Gene Name
thrombospondin 2
Gene Aliases
TSP2
Chromosome Location
Chr.6: 169215780 - 169254114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003247.3 NP_003238.2 13 - 14 2250 62
GenBank mRNA AK292429.1 - 13 - 14 2207 62
BC146676.1 - 12 - 13 2057 62
BC150175.1 - 12 - 13 2061 62
L12350.1 - 13 - 14 2169 62
M81339.1 - 2 - 3 254 62

View Full Product Details