Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THBS2
Entrez Gene ID
7058
Gene Name
thrombospondin 2
Gene Aliases
TSP2
Chromosome Location
Chr.6: 169215780 - 169254114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003247.3 NP_003238.2 15 - 16 2583 91
GenBank mRNA AK292429.1 - 15 - 16 2540 91
BC146676.1 - 14 - 15 2390 91
BC150175.1 - 14 - 15 2394 91
BX641023.1 - 1 - 2 38 91
L12350.1 - 15 - 16 2502 91
M81339.1 - 4 - 5 587 91

View Full Product Details