Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THBS2
Entrez Gene ID
7058
Gene Name
thrombospondin 2
Gene Aliases
TSP2
Chromosome Location
Chr.6: 169215780 - 169254114 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003247.3 NP_003238.2 19 - 20 3327 85
GenBank mRNA AK292429.1 - 19 - 20 3284 85
BC146676.1 - 18 - 19 3134 85
BC150175.1 - 18 - 19 3138 85
BX641023.1 - 5 - 6 782 85
L12350.1 - 19 - 20 3246 85
M81339.1 - 8 - 9 1331 85

View Full Product Details