The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033047.2 | NP_001028219.1 | 7 - 8 | 968 | 105 |
NM_001184690.1 | NP_001171619.1 | 8 - 9 | 1019 | 105 | |
NM_001184691.1 | NP_001171620.1 | 8 - 9 | 1058 | 105 | |
NM_001184692.1 | NP_001171621.1 | 7 - 8 | 968 | 105 | |
NM_001184693.1 | NP_001171622.1 | 8 - 9 | 1058 | 105 | |
XM_005262888.4 | XP_005262945.1 | 9 - 10 | 1096 | 105 | |
XM_005262890.1 | XP_005262947.1 | 7 - 8 | 743 | 105 | |
XM_011531820.2 | XP_011530122.1 | 9 - 10 | 1099 | 105 | |
XM_011531822.2 | XP_011530124.1 | 8 - 9 | 1048 | 105 | |
XM_011531823.2 | XP_011530125.1 | 9 - 10 | 1099 | 105 | |
XM_011531824.2 | XP_011530126.1 | 8 - 9 | 1009 | 105 | |
XM_011531825.2 | XP_011530127.1 | 7 - 8 | 958 | 105 | |
XM_017007984.1 | XP_016863473.1 | 9 - 10 | 1099 | 105 | |
GenBank mRNA | AK290029.1 | - | 7 - 8 | 915 | 105 |
AK295772.1 | - | 8 - 9 | 1019 | 105 | |
AK302477.1 | - | 8 - 9 | 1057 | 105 | |
AK304279.1 | - | 7 - 8 | 967 | 105 | |
AK304721.1 | - | 8 - 9 | 1005 | 105 | |
AL832465.1 | - | 7 - 8 | 882 | 105 | |
AW607534.1 | - | 2 - 3 | 265 | 105 | |
AY358336.1 | - | 7 - 8 | 957 | 105 | |
BC131587.1 | - | 7 - 8 | 815 | 105 |