The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007167.3 | NP_009098.3 | 4 - 5 | 652 | 109 |
GenBank mRNA | AK299293.1 | - | 4 - 5 | 611 | 109 |
AK301461.1 | - | 3 - 4 | 420 | 109 | |
AK310703.1 | - | 4 - 5 | 593 | 109 | |
AL389985.1 | - | 4 - 5 | 577 | 109 | |
BC107880.1 | - | 4 - 5 | 633 | 109 |