Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SIK2
Entrez Gene ID
23235
Gene Name
salt inducible kinase 2
Gene Aliases
LOH11CR1I, QIK, SIK-2, SNF1LK2
Chromosome Location
Chr.11: 111602391 - 111726917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015191.2 NP_056006.1 6 - 7 897 116
XM_017017417.1 XP_016872906.1 6 - 7 856 116
XM_017017418.1 XP_016872907.1 4 - 5 426 116
GenBank mRNA AB018324.3 - 6 - 7 798 116
AB084424.1 - 6 - 7 718 116
AB096248.1 - 6 - 7 803 116
AK000396.1 - 6 - 7 803 116
AK291233.1 - 6 - 7 832 116
AL136764.2 - 6 - 7 820 116
BC078150.1 - 1 - 2 80 116
BC113459.1 - 6 - 7 754 116
BC117183.1 - 6 - 7 754 116

View Full Product Details