Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALG9
Entrez Gene ID
79796
Gene Name
ALG9, alpha-1,2-mannosyltransferase
Gene Aliases
CDG1L, DIBD1, GIKANIS, LOH11CR1J
Chromosome Location
Chr.11: 111776096 - 111871581 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077690.1 NP_001071158.1 7 - 8 892 114
NM_001077691.1 NP_001071159.1 7 - 8 1169 114
NM_001077692.1 NP_001071160.1 7 - 8 1169 114
NM_024740.2 NP_079016.2 7 - 8 892 114
XM_005277723.4 XP_005277780.1 7 - 8 892 114
XM_005277724.4 XP_005277781.1 7 - 8 892 114
XM_006718913.2 XP_006718976.1 7 - 8 892 114
XM_011542992.1 XP_011541294.1 7 - 8 892 114
XM_017018313.1 XP_016873802.1 7 - 8 892 114
XM_017018314.1 XP_016873803.1 7 - 8 892 114
XM_017018315.1 XP_016873804.1 7 - 8 892 114
XM_017018316.1 XP_016873805.1 7 - 8 1189 114
XM_017018317.1 XP_016873806.1 7 - 8 1189 114
XM_017018318.1 XP_016873807.1 7 - 8 1196 114
XM_017018319.1 XP_016873808.1 7 - 8 1026 114
XM_017018320.1 XP_016873809.1 7 - 8 896 114
XM_017018321.1 XP_016873810.1 7 - 8 1032 114
XM_017018322.1 XP_016873811.1 7 - 8 805 114
XM_017018323.1 XP_016873812.1 7 - 8 1189 114
XM_017018324.1 XP_016873813.1 7 - 8 1176 114
XM_017018325.1 XP_016873814.1 8 - 9 1332 114
XM_017018326.1 XP_016873815.1 7 - 8 1193 114
GenBank mRNA AF395532.1 - 7 - 8 844 114
AF454937.1 - 7 - 8 803 114
AK025498.1 - 7 - 8 892 114
AK172828.1 - 7 - 8 753 114
AK225261.1 - 7 - 8 871 114
AK298811.1 - 6 - 7 732 114
AK308025.1 - 6 - 7 787 114
AL136927.1 - 7 - 8 815 114
BC009255.2 - 7 - 8 841 114
KU178783.1 - 7 - 8 794 114
KU178784.1 - 7 - 8 794 114

View Full Product Details