The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016248.3 | NP_057332.1 | 5 - 6 | 383 | 113 |
XM_005266247.1 | XP_005266304.1 | 5 - 6 | 384 | 113 | |
XM_005266249.3 | XP_005266306.1 | 5 - 6 | 384 | 113 | |
XM_005266250.3 | XP_005266307.1 | 5 - 6 | 384 | 113 | |
XM_011534903.2 | XP_011533205.1 | 5 - 6 | 380 | 113 | |
XM_011534904.1 | XP_011533206.1 | 5 - 6 | 576 | 113 | |
XM_011534905.1 | XP_011533207.1 | 4 - 5 | 334 | 113 | |
XM_011534906.2 | XP_011533208.1 | 4 - 5 | 552 | 113 | |
XM_017020381.1 | XP_016875870.1 | 5 - 6 | 434 | 113 | |
GenBank mRNA | AF176555.1 | - | 5 - 6 | 383 | 113 |
CR627087.1 | - | 4 - 5 | 546 | 113 |