The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016248.3 | NP_057332.1 | 9 - 10 | 5453 | 79 |
XM_005266248.3 | XP_005266305.1 | 8 - 9 | 5347 | 79 | |
XM_005266249.3 | XP_005266306.1 | 9 - 10 | 5454 | 79 | |
XM_011534903.2 | XP_011533205.1 | 9 - 10 | 5450 | 79 | |
XM_011534904.1 | XP_011533206.1 | 9 - 10 | 5646 | 79 | |
XM_011534905.1 | XP_011533207.1 | 8 - 9 | 5404 | 79 | |
XM_011534906.2 | XP_011533208.1 | 8 - 9 | 5622 | 79 | |
XM_017020381.1 | XP_016875870.1 | 9 - 10 | 5504 | 79 | |
XM_017020382.1 | XP_016875871.1 | 7 - 8 | 5294 | 79 | |
XM_017020383.1 | XP_016875872.1 | 7 - 8 | 5482 | 79 | |
GenBank mRNA | AB014529.2 | - | 2 - 3 | 3393 | 79 |
AF176555.1 | - | 9 - 10 | 5453 | 79 | |
BE694434.1 | - | 2 - 3 | 310 | 79 |