Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- RAD51B
- Entrez Gene ID
-
5890
- Gene Name
- RAD51 paralog B
- Gene Aliases
-
R51H2, RAD51L1, REC2
- Chromosome Location
-
Chr.14: 67819779 - 68683118 on Build GRCh38
View Full Product Details