The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014728.3 | NP_055543.2 | 9 - 10 | 1435 | 70 |
XM_005274632.3 | XP_005274689.2 | 9 - 10 | 1113 | 70 | |
XM_011545613.2 | XP_011543915.1 | 9 - 10 | 1422 | 70 | |
XM_017029983.1 | XP_016885472.1 | 9 - 10 | 1325 | 70 | |
XM_017029984.1 | XP_016885473.1 | 9 - 10 | 1114 | 70 | |
XM_017029985.1 | XP_016885474.1 | 9 - 10 | 1111 | 70 | |
XM_017029986.1 | XP_016885475.1 | 9 - 10 | 1324 | 70 | |
GenBank mRNA | AB002314.3 | - | 9 - 10 | 1435 | 70 |
AK289694.1 | - | 9 - 10 | 1422 | 70 | |
BC113700.1 | - | 9 - 10 | 1020 | 70 | |
BC113702.1 | - | 9 - 10 | 1020 | 70 | |
BC143746.1 | - | 10 - 11 | 1117 | 70 |