The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014728.3 | NP_055543.2 | 2 - 3 | 667 | 60 |
XM_005274632.3 | XP_005274689.2 | 2 - 3 | 345 | 60 | |
XM_011545613.2 | XP_011543915.1 | 2 - 3 | 654 | 60 | |
XM_017029983.1 | XP_016885472.1 | 2 - 3 | 557 | 60 | |
XM_017029984.1 | XP_016885473.1 | 2 - 3 | 346 | 60 | |
XM_017029985.1 | XP_016885474.1 | 2 - 3 | 343 | 60 | |
XM_017029986.1 | XP_016885475.1 | 2 - 3 | 556 | 60 | |
GenBank mRNA | AB002314.3 | - | 2 - 3 | 667 | 60 |
AK289694.1 | - | 2 - 3 | 654 | 60 | |
BC113700.1 | - | 2 - 3 | 252 | 60 | |
BC113702.1 | - | 2 - 3 | 252 | 60 | |
BC143746.1 | - | 3 - 4 | 349 | 60 |