Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD4
Entrez Gene ID
9758
Gene Name
FERM and PDZ domain containing 4
Gene Aliases
PDZD10, PDZK10
Chromosome Location
Chr.X: 12137329 - 12724523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014728.3 NP_055543.2 4 - 5 929 66
XM_005274632.3 XP_005274689.2 4 - 5 607 66
XM_011545613.2 XP_011543915.1 4 - 5 916 66
XM_017029983.1 XP_016885472.1 4 - 5 819 66
XM_017029984.1 XP_016885473.1 4 - 5 608 66
XM_017029985.1 XP_016885474.1 4 - 5 605 66
XM_017029986.1 XP_016885475.1 4 - 5 818 66
GenBank mRNA AB002314.3 - 4 - 5 929 66
AK289694.1 - 4 - 5 916 66
BC113700.1 - 4 - 5 514 66
BC113702.1 - 4 - 5 514 66
BC143746.1 - 5 - 6 611 66

View Full Product Details