The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080407.2 | NP_001073876.2 | 15 - 16 | 1861 | 128 |
XM_011542566.2 | XP_011540868.1 | 14 - 15 | 1931 | 128 | |
XM_011542567.2 | XP_011540869.1 | 14 - 15 | 1931 | 128 | |
XM_011542568.2 | XP_011540870.1 | 14 - 15 | 1895 | 128 | |
XM_011542569.2 | XP_011540871.1 | 10 - 11 | 1520 | 128 | |
XM_011542570.2 | XP_011540872.1 | 10 - 11 | 1484 | 128 | |
XM_011542572.1 | XP_011540874.1 | 10 - 11 | 1079 | 128 | |
XM_011542573.1 | XP_011540875.1 | 8 - 9 | 1015 | 128 | |
XM_011542574.2 | XP_011540876.1 | 8 - 9 | 1138 | 128 | |
XM_017017155.1 | XP_016872644.1 | 15 - 16 | 2759 | 128 | |
XM_017017156.1 | XP_016872645.1 | 11 - 12 | 2384 | 128 | |
GenBank mRNA | AK074712.1 | - | 6 - 7 | 567 | 128 |