The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153365.2 | NP_699196.2 | 6 - 7 | 916 | 146 |
XM_005248139.2 | XP_005248196.1 | 5 - 6 | 730 | 146 | |
XM_005248140.3 | XP_005248197.1 | 5 - 6 | 680 | 146 | |
XM_011513815.2 | XP_011512117.1 | 5 - 6 | 2015 | 146 | |
XM_011513816.2 | XP_011512118.1 | 5 - 6 | 724 | 146 | |
XM_011513817.2 | XP_011512119.1 | 3 - 4 | 293 | 146 | |
XM_017007876.1 | XP_016863365.1 | 7 - 8 | 806 | 146 | |
GenBank mRNA | AK074494.1 | - | 6 - 7 | 849 | 146 |
AK296104.1 | - | 5 - 6 | 718 | 146 | |
BC066899.1 | - | 6 - 7 | 854 | 146 |