Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAPT1
Entrez Gene ID
202018
Gene Name
transmembrane anterior posterior transformation 1
Gene Aliases
CMVFR, OCLSBG
Chromosome Location
Chr.4: 16160505 - 16226538 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017007876.1 XP_016863365.1 4 - 5 417 86

View Full Product Details