Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC13
Entrez Gene ID
152206
Gene Name
coiled-coil domain containing 13
Gene Aliases
-
Chromosome Location
Chr.3: 42704118 - 42773253 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144719.3 NP_653320.3 7 - 8 911 69
XM_005264898.3 XP_005264955.1 7 - 8 911 69
XM_005264899.4 XP_005264956.1 7 - 8 911 69
XM_011533418.2 XP_011531720.1 7 - 8 911 69
XM_011533419.2 XP_011531721.1 3 - 4 922 69
XM_011533420.2 XP_011531722.1 7 - 8 911 69
XM_011533421.2 XP_011531723.1 7 - 8 911 69
XM_017005781.1 XP_016861270.1 3 - 4 309 69
GenBank mRNA BC036050.1 - 7 - 8 913 69

View Full Product Details