The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317784.1 | NP_001304713.1 | 10 - 11 | 1679 | 154 |
NM_017988.5 | NP_060458.3 | 10 - 11 | 1831 | 154 | |
XM_017019618.1 | XP_016875107.1 | 11 - 12 | 1691 | 154 | |
XM_017019619.1 | XP_016875108.1 | 11 - 12 | 1843 | 154 | |
XM_017019620.1 | XP_016875109.1 | 12 - 13 | 1830 | 154 | |
GenBank mRNA | AB037781.1 | - | 8 - 9 | 997 | 154 |
AK024274.1 | - | 6 - 7 | 840 | 154 | |
AK125091.1 | - | 10 - 11 | 1816 | 154 | |
AK292980.1 | - | 10 - 11 | 1605 | 154 | |
BC012387.2 | - | 1 - 2 | 124 | 154 | |
BC063798.1 | - | 10 - 11 | 1444 | 154 |