The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005708.3 | NP_005699.1 | 5 - 6 | 1641 | 117 |
XM_011521044.2 | XP_011519346.1 | 5 - 6 | 966 | 117 | |
XM_017020298.1 | XP_016875787.1 | 5 - 6 | 3249 | 117 | |
XM_017020299.1 | XP_016875788.1 | 5 - 6 | 2016 | 117 | |
XM_017020300.1 | XP_016875789.1 | 5 - 6 | 873 | 117 | |
XM_017020301.1 | XP_016875790.1 | 4 - 5 | 844 | 117 | |
XM_017020302.1 | XP_016875791.1 | 2 - 3 | 638 | 117 | |
GenBank mRNA | AF105267.1 | - | 5 - 6 | 1594 | 117 |
AF111178.1 | - | 5 - 6 | 1643 | 117 | |
AK074741.1 | - | 6 - 7 | 1095 | 117 | |
AK290144.1 | - | 5 - 6 | 1162 | 117 | |
AK304338.1 | - | 5 - 6 | 1460 | 117 | |
AY358462.1 | - | 5 - 6 | 1574 | 117 | |
BC106947.2 | - | 5 - 6 | 1489 | 117 |