Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CCDC171
- Entrez Gene ID
-
203238
- Gene Name
- coiled-coil domain containing 171
- Gene Aliases
-
C9orf93, bA536D16.1, bA778P13.1
- Chromosome Location
-
Chr.9: 15552874 - 16062431 on Build GRCh38
View Full Product Details