Gene Symbol | Transcript Accession | Exon Location | Assay Location | Protein ID |
---|---|---|---|---|
RECQL4 | NM_004260.3 | NP_004251.3 | ||
XM_011517384.2 | XP_011515686.1 | |||
XM_017013991.1 | XP_016869480.1 | |||
XM_017013992.1 | XP_016869481.1 | |||
XM_017013993.1 | XP_016869482.1 | |||
XM_017013994.1 | XP_016869483.1 | |||
XM_017013995.1 | XP_016869484.1 | |||
XM_017013996.1 | XP_016869485.1 | |||
XM_017013997.1 | XP_016869486.1 | |||
XM_017013998.1 | XP_016869487.1 | |||
XM_017013999.1 | XP_016869488.1 | |||
XM_017014000.1 | XP_016869489.1 | |||
XM_017014001.1 | XP_016869490.1 | |||
AB006532.1 | BAA74453.1 | |||
KJ147518.1 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv470249 | Chr8:143889098 - 144519234 on Build GRCh38 | Loss | SMPD5 FOXH1 CYHR1 DGAT1 MIR1234 SHARPIN SLC52A2 PLEC MAF1 TMEM249 PPP1R16A BOP1 EXOSC4 GPAA1 MFSD3 SCX KIFC2 HGH1 MIR6846 LOC101928953 ADCK5 RECQL4 OPLAH LRRC14 MIR6893 WDR97 MIR7112 LOC101928902 FBXL6 CYC1 MIR6849 TONSL TONSL-AS1 HSF1 CPSF1 MIR6847 GRINA SCRT1 MIR939 SPATC1 MROH1 GPT PARP10 VPS28 MIR6848 MIR661 SLC39A4 |
nsv612951 | Chr8:144479830 - 144677692 on Build GRCh38 | Loss | MFSD3 LOC101928953 RECQL4 GPT LRRC24 LRRC14 C8orf82 PPP1R16A ARHGAP39 |
nsv831489 | Chr8:144342140 - 144544026 on Build GRCh38 | Loss | FOXH1 CYHR1 MIR1234 SLC52A2 TMEM249 PPP1R16A ARHGAP39 MFSD3 KIFC2 LOC101928953 ADCK5 RECQL4 LRRC14 MIR6893 LOC101928902 FBXL6 MIR6849 TONSL TONSL-AS1 C8orf82 CPSF1 MIR939 GPT LRRC24 VPS28 SLC39A4 |
dgv1207n67 | Chr8:144498396 - 144532417 on Build GRCh38 | Gain | MFSD3 LOC101928953 RECQL4 GPT LRRC24 LRRC14 C8orf82 PPP1R16A ARHGAP39 |
dgv12439n54 | Chr8:144438306 - 144525122 on Build GRCh38 | Loss | FOXH1 CYHR1 MFSD3 KIFC2 LOC101928953 RECQL4 TONSL TONSL-AS1 GPT LRRC24 LRRC14 PPP1R16A |
dgv907n27 | Chr8:144416180 - 144525122 on Build GRCh38 | Loss | FOXH1 CYHR1 TONSL TONSL-AS1 PPP1R16A MFSD3 KIFC2 LOC101928953 RECQL4 GPT LRRC24 LRRC14 MIR6893 VPS28 SLC39A4 |
dgv12442n54 | Chr8:144492696 - 144516381 on Build GRCh38 | Loss | MFSD3 LOC101928953 RECQL4 GPT PPP1R16A |
dgv12434n54 | Chr8:144270080 - 144516801 on Build GRCh38 | Loss | FOXH1 CYHR1 DGAT1 MIR1234 SLC52A2 TMEM249 PPP1R16A BOP1 MFSD3 KIFC2 LOC101928953 ADCK5 RECQL4 MIR6893 LOC101928902 FBXL6 MIR6849 TONSL TONSL-AS1 HSF1 CPSF1 SCRT1 MIR939 GPT VPS28 MIR6848 SLC39A4 |
dgv12443n54 | Chr8:144502322 - 144518490 on Build GRCh38 | Loss | MFSD3 LOC101928953 RECQL4 GPT LRRC14 |
nsv515809 | Chr8:143904439 - 144519234 on Build GRCh38 | Gain+Loss | SMPD5 FOXH1 CYHR1 DGAT1 MIR1234 SHARPIN SLC52A2 PLEC MAF1 TMEM249 PPP1R16A BOP1 EXOSC4 GPAA1 MFSD3 SCX KIFC2 HGH1 MIR6846 LOC101928953 ADCK5 RECQL4 OPLAH LRRC14 MIR6893 WDR97 MIR7112 LOC101928902 FBXL6 CYC1 MIR6849 TONSL TONSL-AS1 HSF1 CPSF1 MIR6847 GRINA SCRT1 MIR939 SPATC1 MROH1 GPT PARP10 VPS28 MIR6848 MIR661 SLC39A4 |
nsv612940 | Chr8:144436292 - 144613673 on Build GRCh38 | Loss | FOXH1 CYHR1 TONSL TONSL-AS1 C8orf82 PPP1R16A ARHGAP39 MFSD3 KIFC2 LOC101928953 RECQL4 GPT LRRC24 LRRC14 |
nsv428208 | Chr8:144041826 - 144722291 on Build GRCh38 | Gain | SMPD5 FOXH1 CYHR1 DGAT1 MIR1234 SHARPIN SLC52A2 MAF1 TMEM249 PPP1R16A ARHGAP39 BOP1 EXOSC4 GPAA1 MFSD3 SCX KIFC2 HGH1 MIR6846 LOC101928953 ADCK5 RECQL4 OPLAH LRRC14 MIR6893 WDR97 MIR7112 LOC101928902 FBXL6 CYC1 MIR6849 TONSL TONSL-AS1 HSF1 C8orf82 CPSF1 MIR6847 SCRT1 MIR939 SPATC1 MROH1 GPT LRRC24 ZNF251 VPS28 MIR6848 SLC39A4 |
nsv612965 | Chr8:144508945 - 144516381 on Build GRCh38 | Loss | MFSD3 RECQL4 |
esv29999 | Chr8:144456732 - 144525122 on Build GRCh38 | Loss | FOXH1 CYHR1 MFSD3 KIFC2 LOC101928953 RECQL4 GPT LRRC24 LRRC14 PPP1R16A |