Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SNRNP48
- Entrez Gene ID
-
154007
- Gene Name
- small nuclear ribonucleoprotein U11/U12 subunit 48
- Gene Aliases
-
C6orf151, dJ336K20B.1, dJ512B11.2
- Chromosome Location
-
Chr.6: 7590199 - 7611967 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK056796.1 |
- |
4 - 5 |
442 |
81 |
View Full Product Details