The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001284274.1 | NP_001271203.1 | 10 - 11 | 1142 | 156 |
NM_182765.4 | NP_877497.3 | 9 - 10 | 1130 | 156 | |
XM_017015750.1 | XP_016871239.1 | 9 - 10 | 980 | 156 | |
XM_017015751.1 | XP_016871240.1 | 9 - 10 | 1076 | 156 | |
XM_017015752.1 | XP_016871241.1 | 2 - 3 | 334 | 156 | |
GenBank mRNA | AK295723.1 | - | 10 - 11 | 1085 | 156 |
AL832073.1 | - | 1 - 2 | 572 | 156 | |
BC040187.1 | - | 9 - 10 | 1073 | 156 |