The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291285.1 | NP_001278214.1 | 14 - 15 | 12607 | 73 |
NM_001291303.1 | NP_001278232.1 | 14 - 15 | 12607 | 73 | |
NM_024582.4 | NP_078858.4 | 14 - 15 | 12601 | 73 | |
XM_011532236.2 | XP_011530538.1 | 15 - 16 | 13684 | 73 | |
XM_011532237.2 | XP_011530539.1 | 14 - 15 | 8492 | 73 | |
GenBank mRNA | AK096646.1 | - | 6 - 7 | 903 | 73 |
AK291461.1 | - | 6 - 7 | 1147 | 73 |