The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 7 - 8 | 1170 | 126 |
XM_011542067.2 | XP_011540369.1 | 8 - 9 | 2102 | 126 | |
XM_011542068.2 | XP_011540370.1 | 8 - 9 | 2107 | 126 | |
XM_011542069.2 | XP_011540371.1 | 7 - 8 | 1961 | 126 | |
XM_011542070.2 | XP_011540372.1 | 6 - 7 | 1128 | 126 | |
XM_011542071.2 | XP_011540373.1 | 5 - 6 | 1537 | 126 | |
XM_017002225.1 | XP_016857714.1 | 7 - 8 | 1187 | 126 | |
XM_017002226.1 | XP_016857715.1 | 8 - 9 | 1498 | 126 | |
GenBank mRNA | AL832217.1 | - | 14 - 15 | 2203 | 126 |
AY639646.1 | - | 6 - 7 | 995 | 126 | |
BX640642.1 | - | 3 - 4 | 414 | 126 |