The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080850.2 | NP_001074319.1 | 9 - 10 | 1489 | 78 |
XM_011542067.2 | XP_011540369.1 | 10 - 11 | 2421 | 78 | |
XM_011542068.2 | XP_011540370.1 | 10 - 11 | 2426 | 78 | |
XM_011542069.2 | XP_011540371.1 | 9 - 10 | 2280 | 78 | |
XM_011542070.2 | XP_011540372.1 | 8 - 9 | 1447 | 78 | |
XM_011542071.2 | XP_011540373.1 | 7 - 8 | 1856 | 78 | |
XM_017002225.1 | XP_016857714.1 | 9 - 10 | 1506 | 78 | |
XM_017002226.1 | XP_016857715.1 | 10 - 11 | 1817 | 78 | |
GenBank mRNA | AL832217.1 | - | 16 - 17 | 2522 | 78 |
AY639646.1 | - | 8 - 9 | 1314 | 78 |