Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5orf58
Entrez Gene ID
133874
Gene Name
chromosome 5 open reading frame 58
Gene Aliases
-
Chromosome Location
Chr.5: 170231730 - 170252569 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_131091.1 3 - 4 276 162
GenBank mRNA BC030767.1 - 3 - 4 256 162

View Full Product Details