Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHGB3
Entrez Gene ID
56102
Gene Name
protocadherin gamma subfamily B, 3
Gene Aliases
PCDH-GAMMA-B3
Chromosome Location
Chr.5: 141370264 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018924.4 NP_061747.2 1 - 1 2460 95
NM_032097.2 NP_115268.2 1 - 1 2460 95
GenBank mRNA AF152332.1 - 1 - 1 2329 95
AF152519.1 - 1 - 1 2329 95
BC150172.1 - 1 - 1 2460 95

View Full Product Details