The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032053.2 | NP_114442.2 | 1 - 1 | 2627 | 86 |
GenBank mRNA | AI805761.1 | - | 1 - 1 | 86 | |
BX100615.1 | - | 1 - 1 | 86 |