Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCDHGA1
Entrez Gene ID
56114
Gene Name
protocadherin gamma subfamily A, 1
Gene Aliases
PCDH-GAMMA-A1
Chromosome Location
Chr.5: 141330685 - 141512979 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031993.1 NP_114382.1 1 - 1 2419 92
GenBank mRNA AF152507.1 - 1 - 1 2419 92
BC112082.1 - 1 - 1 2467 92
BC112084.1 - 1 - 1 2467 92

View Full Product Details