The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052900.2 | NP_443132.3 | 27 - 28 | 4601 | 119 |
NM_198123.1 | NP_937756.1 | 28 - 29 | 4913 | 119 | |
NM_198124.1 | NP_937757.1 | 29 - 30 | 4805 | 119 | |
XM_011516815.2 | XP_011515117.1 | 2 - 3 | 414 | 119 | |
XM_011516816.2 | XP_011515118.1 | 27 - 28 | 5087 | 119 | |
XM_017013008.1 | XP_016868497.1 | 27 - 28 | 5321 | 119 | |
XM_017013009.1 | XP_016868498.1 | 28 - 29 | 5399 | 119 | |
XM_017013010.1 | XP_016868499.1 | 26 - 27 | 5009 | 119 | |
XM_017013011.1 | XP_016868500.1 | 26 - 27 | 5009 | 119 | |
XM_017013012.1 | XP_016868501.1 | 26 - 27 | 4372 | 119 | |
GenBank mRNA | AB067481.2 | - | 15 - 16 | 2774 | 119 |
AB114604.1 | - | 28 - 29 | 4913 | 119 | |
AB114605.1 | - | 29 - 30 | 4805 | 119 | |
AY210419.1 | - | 27 - 28 | 4472 | 119 |