Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSMD3
Entrez Gene ID
114788
Gene Name
CUB and Sushi multiple domains 3
Gene Aliases
-
Chromosome Location
Chr.8: 112222928 - 113437499 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052900.2 NP_443132.3 48 - 49 7536 114
NM_198123.1 NP_937756.1 49 - 50 7848 114
NM_198124.1 NP_937757.1 50 - 51 7740 114
XM_011516815.2 XP_011515117.1 23 - 24 3349 114
XM_011516816.2 XP_011515118.1 48 - 49 8022 114
XM_017013008.1 XP_016868497.1 48 - 49 8256 114
XM_017013009.1 XP_016868498.1 47 - 48 8124 114
XM_017013010.1 XP_016868499.1 47 - 48 7944 114
XM_017013011.1 XP_016868500.1 45 - 46 7734 114
XM_017013012.1 XP_016868501.1 47 - 48 7307 114
GenBank mRNA AB067481.2 - 34 - 35 5499 114
AB114604.1 - 49 - 50 7848 114
AB114605.1 - 50 - 51 7740 114
AK126252.1 - 7 - 8 881 114
AY210419.1 - 48 - 49 7407 114

View Full Product Details