Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
-
Chromosome Location
Chr.9: 113410644 - 113429684 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278629.1 NP_001265558.1 1 - 2 361 75
XM_011518477.1 XP_011516779.1 1 - 2 361 75
XM_011518478.1 XP_011516780.1 1 - 2 361 75
GenBank mRNA AK313867.1 - 1 - 2 350 75
BC033037.2 - 1 - 2 319 75
DB057586.1 - 1 - 2 362 75

View Full Product Details