The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174882.2 | NP_777378.1 | 1 - 2 | 437 | 74 |
GenBank mRNA | AY103469.1 | - | 1 - 2 | 437 | 75 |