Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM208B
Entrez Gene ID
54906
Gene Name
family with sequence similarity 208 member B
Gene Aliases
C10orf18, bA318E3.2
Chromosome Location
Chr.10: 5684759 - 5763740 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321783.1 NP_001308712.1 14 - 15 3021 80
NM_001321784.1 NP_001308713.1 14 - 15 3067 80
NM_001321785.1 NP_001308714.1 13 - 14 2778 80
NM_017782.4 NP_060252.4 13 - 14 2948 80
XM_005252474.3 XP_005252531.1 16 - 17 9759 80
XM_005252475.3 XP_005252532.1 17 - 18 3420 80
XM_005252476.4 XP_005252533.1 15 - 16 3203 80
XM_005252480.4 XP_005252537.1 13 - 14 3008 80
XM_005252481.4 XP_005252538.1 11 - 12 2493 80
XM_005252482.2 XP_005252539.1 15 - 16 2853 80
XM_011519515.2 XP_011517817.1 17 - 18 9897 80
XM_011519516.2 XP_011517818.1 17 - 18 9897 80
XM_011519517.2 XP_011517819.1 17 - 18 3817 80
XM_011519518.2 XP_011517820.1 16 - 17 3130 80
XM_011519519.2 XP_011517821.1 16 - 17 9654 80
XM_011519520.2 XP_011517822.1 14 - 15 9594 80
XM_011519521.2 XP_011517823.1 16 - 17 3341 80
XM_011519522.2 XP_011517824.1 16 - 17 3725 80
XM_011519525.2 XP_011517827.1 14 - 15 2761 80
XM_017016361.1 XP_016871850.1 16 - 17 9654 80
XM_017016362.1 XP_016871851.1 15 - 16 3624 80
XM_017016363.1 XP_016871852.1 16 - 17 3282 80
XM_017016364.1 XP_016871853.1 14 - 15 3486 80
XM_017016365.1 XP_016871854.1 15 - 16 3148 80
GenBank mRNA AK128051.1 - 1 - 2 223 80
BX649177.1 - 5 - 6 1921 80
CR936780.1 - 13 - 14 2949 80

View Full Product Details