The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020700.1 | NP_065751.1 | 5 - 6 | 1105 | 62 |
XM_011538578.2 | XP_011536880.1 | 5 - 6 | 1241 | 62 | |
XM_011538579.1 | XP_011536881.1 | 5 - 6 | 793 | 62 | |
XM_017019676.1 | XP_016875165.1 | 5 - 6 | 1355 | 62 | |
GenBank mRNA | AB032983.1 | - | 4 - 5 | 698 | 62 |
AB084258.1 | - | 5 - 6 | 1370 | 62 | |
AK023315.1 | - | 6 - 7 | 1167 | 62 | |
BC157843.1 | - | 5 - 6 | 1115 | 62 |