Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADCYAP1
Entrez Gene ID
116
Gene Name
adenylate cyclase activating polypeptide 1
Gene Aliases
PACAP
Chromosome Location
Chr.18: 904342 - 912172 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099733.1 NP_001093203.1 3 - 4 363 137
NM_001117.4 NP_001108.2 2 - 3 435 137
XM_005258081.3 XP_005258138.1 4 - 5 762 137
GenBank mRNA AK294233.1 - 4 - 5 691 137
AK313050.1 - 3 - 4 363 137
BC093837.1 - 2 - 3 315 137
BC101803.1 - 2 - 3 315 137
DC320719.1 - 3 - 4 363 137
DN991692.1 - 2 - 3 342 137
S83513.1 - 2 - 3 245 137

View Full Product Details