Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSLP
Entrez Gene ID
85480
Gene Name
thymic stromal lymphopoietin
Gene Aliases
-
Chromosome Location
Chr.5: 111070080 - 111078024 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033035.4 NP_149024.1 2 - 3 420 69
NR_045089.1 3 - 4 1625 69
GenBank mRNA AF338732.1 - 2 - 3 254 69
AY037115.1 - 2 - 3 420 69
BC040592.1 - 3 - 4 1626 69

View Full Product Details