The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001785.2 | NP_001776.1 | 3 - 4 | 510 | 62 |
GenBank mRNA | AJ000474.1 | - | 3 - 4 | 337 | 62 |
BC048284.1 | - | 4 - 5 | 633 | 62 | |
BC054036.1 | - | 3 - 4 | 395 | 62 | |
CD675458.1 | - | 3 - 4 | 498 | 62 | |
L27943.1 | - | 3 - 4 | 448 | 62 | |
S52873.1 | - | 3 - 4 | 328 | 62 |