Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFAP2C
Entrez Gene ID
7022
Gene Name
transcription factor AP-2 gamma
Gene Aliases
AP2-GAMMA, ERF1, TFAP2G, hAP-2g
Chromosome Location
Chr.20: 56629302 - 56639283 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003222.3 NP_003213.1 2 - 3 776 70
GenBank mRNA AK301572.1 - 2 - 3 592 70
AK314272.1 - 2 - 3 772 70
BC035664.1 - 2 - 3 735 70
BC051829.1 - 2 - 3 776 70
U85658.1 - 2 - 3 699 70
X95693.1 - 2 - 3 579 70

View Full Product Details